Merck
  • Elastic, Persistently Moisture-Retentive, and Wearable Biomimetic Film Inspired by Fetal Scarless Repair for Promoting Skin Wound Healing.
Elastic, Persistently Moisture-Retentive, and Wearable Biomimetic Film Inspired by Fetal Scarless Repair for Promoting Skin Wound Healing.
ACS applied materials & interfaces (2020-01-16)
Dongmei Zhang, Guanke Cai, Somnath Mukherjee, Yajuan Sun, Changhao Wang, Bingjie Mai, Kaiqiang Liu, Cheng Yang, Yashao Chen
ABSTRACT
An efficient and available material for promoting skin regeneration is of great importance for public health, but it remains an elusive goal. Inspired by fetal scarless wound healing, we develop a wearable biomimetic film (WBMF) composed of hyaluronan (HA), vitamin E (VE), dopamine (DA), and β-cyclodextrin (β-CD) that mimics the fetal context (FC) and fetal extracellular matrix (ECM) around the wound bed for dermal regeneration. First, the WBMF creates the FC of sterility, hypoxia, persistent moisture, and no secondary insults for wounds as the result of its seamless adhesion to the skin, optimum stress-stretch and high-cycle fatigue resistance matching the anisotropic tension of the skin, and water-triggered self-healing behavior. Thus, the WBMF modulates the early wound situation to minimize inflammatory response. In the meantime, the WBMF mimics the critical biological function of fetal ECM, inducing fibroblast migration, suppressing the overexpression of transforming growth factor β1, and mediating collagen synthesis, distribution, and reestablishment. As a result, the WBMF accelerates wound healing and gains a normal dermal collagen architecture, thereby restoring scarless appearance. Overall, the WBMF provides a new paradigm for promoting skin wound healing and may find broad utility for the field of regenerative medicine.
Sigma-Aldrich
4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium tetrafluoroborate, 97%