Merck
  • NFIA and GATA3 are crucial regulators of embryonic articular cartilage differentiation.
NFIA and GATA3 are crucial regulators of embryonic articular cartilage differentiation.
Development (Cambridge, England) (2017-12-17)
Pratik Narendra Pratap Singh, Upendra Singh Yadav, Kimi Azad, Pooja Goswami, Veena Kinare, Amitabha Bandyopadhyay
ABSTRACT
During appendicular skeletal development, the bi-potential cartilage anlagen gives rise to transient cartilage, which is eventually replaced by bone, and to articular cartilage that caps the ends of individual skeletal elements. While the molecular mechanism that regulates transient cartilage differentiation is relatively well understood, the mechanism of articular cartilage differentiation has only begun to be unraveled. Furthermore, the molecules that coordinate the articular and transient cartilage differentiation processes are poorly understood. Here, we have characterized in chick the regulatory roles of two transcription factors, NFIA and GATA3, in articular cartilage differentiation, maintenance and the coordinated differentiation of articular and transient cartilage. Both NFIA and GATA3 block hypertrophic differentiation. Our results suggest that NFIA is not sufficient but necessary for articular cartilage differentiation. Ectopic activation of GATA3 promotes articular cartilage differentiation, whereas inhibition of GATA3 activity promotes transient cartilage differentiation at the expense of articular cartilage. We propose a novel transcriptional circuitry involved in embryonic articular cartilage differentiation, maintenance and its crosstalk with the transient cartilage differentiation program.