Merck
  • The ubiquitin-specific protease USP8 directly deubiquitinates SQSTM1/p62 to suppress its autophagic activity.
The ubiquitin-specific protease USP8 directly deubiquitinates SQSTM1/p62 to suppress its autophagic activity.
Autophagy (2019-06-27)
Hong Peng, Fang Yang, Qianwen Hu, Jian Sun, Cheng Peng, Yao Zhao, Chuanxin Huang
ABSTRACT
SQSTM1/p62 (sequestosome 1) is a critical macroautophagy/autophagy receptor that promotes the formation and degradation of ubiquitinated aggregates. SQSTM1 can be modified by ubiquitination, and this modification modulates its autophagic activity. However, the molecular mechanisms underpinning its reversible deubiquitination have never been described. Here we report that USP8 (ubiquitin specific peptidase 8) directly interacted with and deubiquitinated SQSTM1. USP8 preferentially removed the lysine 11 (K11)-linked ubiquitin chains from SQSTM1. Moreover, USP8 deubiquitinated SQSTM1 principally at K420 within its ubiquitin-association (UBA) domain. Finally, USP8 inhibited SQSTM1 degradation and autophagic influx in cells with wild-type SQSTM1, but not its mutant with substitution of K420 with an arginine. Taken together, USP8 acts as a negative regulator of autophagy by deubiquitinating SQSTM1 at K420.Abbreviations: BafA1: bafilomycin A1; BAP1: BRCA1 associated protein 1; DUB: deubiquitinating enzyme; ESCRT: endosomal sorting complex required for transport; HTT: huntingtin; K: lysine; KEAP1: kelch like ECH associated protein 1; MAP1LC3/LC3: microtubule associated protein 1 light chain 3; MEF: mouse embryonic fibroblast; shRNA: short hairpin RNA; SQSTM1: sequestosome 1; Ub: ubiquitin; UBA: ubiquitin-association; UBE2D2: ubiquitin conjugating enzyme E2 D2; UBE2D3: ubiquitin conjugating enzyme E2 D3; USP: ubiquitin specific peptidase; WT: wild-type.
Sigma-Aldrich
3X FLAG® Peptide, lyophilized powder
Sigma-Aldrich
MISSION® esiRNA, targeting human USP8
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Sigma-Aldrich
Triton X-100, laboratory grade
Roche
cOmplete, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks