Merck
  • Pattern recognition receptor-mediated innate immune responses in seminal vesicle epithelial cell and their impacts on cellular function†.
Pattern recognition receptor-mediated innate immune responses in seminal vesicle epithelial cell and their impacts on cellular function†.
Biology of reproduction (2019-07-28)
Maolei Gong, Fei Wang, Weihua Liu, Ran Chen, Han Wu, Wenjing Zhang, Xiaoqin Yu, Ruiqin Han, Aijie Liu, Yongmei Chen, Daishu Han
ABSTRACT
The seminal vesicles can be infected by microorganisms, thereby resulting in vesiculitis and impairment in male fertility. Innate immune responses in seminal vesicles cells to microbial infections, which facilitate vesiculitis, have yet to be investigated. The present study aims to elucidate pattern recognition receptor-mediated innate immune responses in seminal vesicles epithelial cells. Various pattern recognition receptors, including Toll-like receptor 3, Toll-like receptor 4, cytosolic ribonucleic acid, and deoxyribonucleic acid sensors, are abundantly expressed in seminal vesicles epithelial cells. These pattern recognition receptors can recognize their respective ligands, thus activating nuclear factor kappa B and interferon regulatory factor 3. The pattern recognition receptor signaling induces expression of pro-inflammatory cytokines, such as tumor necrosis factor alpha (Tnfa) and interleukin 6 (Il6), chemokines monocyte chemoattractant protein-1 (Mcp1) and C-X-C motif chemokine 10 (Cxcl10), and type 1 interferons Ifna and Ifnb. Moreover, pattern recognition receptor-mediated innate immune responses up-regulated the expression of microsomal prostaglandin E synthase and cyclooxygenase 2, but they down-regulated semenogelin-1 expression. These results provide novel insights into the mechanism underlying vesiculitis and its impact on the functions of the seminal vesicles.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by gel-filtration chromatography
Sigma-Aldrich
Collagenase type I, The collagenase type I (from Clostridium histolyticum) is a crude collagenase preparation that can be used for the isolation of primary cells or for tissue dissociation by enzymatic means.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid