Merck
  • Inhibition of vascular calcification by inositol phosphates derivatized with ethylene glycol oligomers.
Inhibition of vascular calcification by inositol phosphates derivatized with ethylene glycol oligomers.
Nature communications (2020-02-07)
Antonia E Schantl, Anja Verhulst, Ellen Neven, Geert J Behets, Patrick C D'Haese, Marc Maillard, David Mordasini, Olivier Phan, Michel Burnier, Dany Spaggiari, Laurent A Decosterd, Mark G MacAskill, Carlos J Alcaide-Corral, Adriana A S Tavares, David E Newby, Victoria C Beindl, Roberto Maj, Anne Labarre, Chrismita Hegde, Bastien Castagner, Mattias E Ivarsson, Jean-Christophe Leroux
ABSTRACT
Myo-inositol hexakisphosphate (IP6) is a natural product known to inhibit vascular calcification (VC), but with limited potency and low plasma exposure following bolus administration. Here we report the design of a series of inositol phosphate analogs as crystallization inhibitors, among which 4,6-di-O-(methoxy-diethyleneglycol)-myo-inositol-1,2,3,5-tetrakis(phosphate), (OEG2)2-IP4, displays increased in vitro activity, as well as more favorable pharmacokinetic and safety profiles than IP6 after subcutaneous injection. (OEG2)2-IP4 potently stabilizes calciprotein particle (CPP) growth, consistently demonstrates low micromolar activity in different in vitro models of VC (i.e., human serum, primary cell cultures, and tissue explants), and largely abolishes the development of VC in rodent models, while not causing toxicity related to serum calcium chelation. The data suggest a mechanism of action independent of the etiology of VC, whereby (OEG2)2-IP4 disrupts the nucleation and growth of pathological calcification.
Product Description
Supelco
Amber storage bottles, volume 240 mL, O.D. × H 61 mm × 143 mm, thread for 24-400, bottle only, pkg of 12 ea
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Millipore
myo-Inositol, ≥99.0%, suitable for microbiology
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Millipore
Magnesium citrate tribasic nonahydrate, suitable for microbiology, ≥95.0% (KT)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Magnesium citrate, AldrichCPR
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Supelco
Sodium dihydrogen phosphate monohydrate, for analysis EMSURE® ACS,Reag. Ph Eur
Sigma-Aldrich
Human Serum, from human male AB plasma, USA origin, sterile-filtered