Merck
  • Synthesis of silver nanoparticles colloids in imidazolium halide ionic liquids and their antibacterial activities for gram-positive and gram-negative bacteria.
Synthesis of silver nanoparticles colloids in imidazolium halide ionic liquids and their antibacterial activities for gram-positive and gram-negative bacteria.
Chemosphere (2019-11-15)
Virendra Patil, Swapnil Mahajan, Mohan Kulkarni, Kashinath Patil, Chandrashekhar Rode, Alberto Coronas, Gi-Ra Yi
ABSTRACT
Four 1-butyl-3-methylimidazolium halide ionic liquids were synthesized via metathesis and anion exchange reactions. Silver nanoparticles (AgNPs) colloids were synthesized in four ionic liquids in the pressurized reactor by reduction of silver nitrate with hydrogen gas, without adding solvents or stabilizing agents. Antibacterial activities of base ionic liquids and AgNPs colloids in ionic liquids were reviewed by well-diffusion method for gram-positive Bacillus cereus (NCIM-2155) and gram-negative Escherichia coli (NCIM-2931) bacteria. Antibacterial activities of ionic liquids and AgNPs colloids in ionic liquids were observed to be controlled by ionic liquids anions and AgNPs particle size. The 1-butyl-3-methylimidazolium iodide ionic liquid exhibited higher antibacterial activities among the studied ionic liquids. Further, the presence of AgNPs in 1-butyl-3-methylimidazolium iodide, ionic liquid enhanced its antibacterial activity for Bacillus cereus and Escherichia coli bacteria.
Sigma-Aldrich
1-Chlorobutane, ReagentPlus®, 99%
Sigma-Aldrich
1-Iodobutane, 99%, contains copper as stabilizer
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)