Merck
  • PRIMPOL-Mediated Adaptive Response Suppresses Replication Fork Reversal in BRCA-Deficient Cells.
PRIMPOL-Mediated Adaptive Response Suppresses Replication Fork Reversal in BRCA-Deficient Cells.
Molecular cell (2019-11-05)
Annabel Quinet, Stephanie Tirman, Jessica Jackson, Saša Šviković, Delphine Lemaçon, Denisse Carvajal-Maldonado, Daniel González-Acosta, Alexandre T Vessoni, Emily Cybulla, Matthew Wood, Steven Tavis, Luis F Z Batista, Juan Méndez, Julian E Sale, Alessandro Vindigni
ABSTRACT
Acute treatment with replication-stalling chemotherapeutics causes reversal of replication forks. BRCA proteins protect reversed forks from nucleolytic degradation, and their loss leads to chemosensitivity. Here, we show that fork degradation is no longer detectable in BRCA1-deficient cancer cells exposed to multiple cisplatin doses, mimicking a clinical treatment regimen. This effect depends on increased expression and chromatin loading of PRIMPOL and is regulated by ATR activity. Electron microscopy and single-molecule DNA fiber analyses reveal that PRIMPOL rescues fork degradation by reinitiating DNA synthesis past DNA lesions. PRIMPOL repriming leads to accumulation of ssDNA gaps while suppressing fork reversal. We propose that cells adapt to repeated cisplatin doses by activating PRIMPOL repriming under conditions that would otherwise promote pathological reversed fork degradation. This effect is generalizable to other conditions of impaired fork reversal (e.g., SMARCAL1 loss or PARP inhibition) and suggests a new strategy to modulate cisplatin chemosensitivity by targeting the PRIMPOL pathway.
Sigma-Aldrich
MISSION® esiRNA, targeting human SMARCAL1
Sigma-Aldrich
MISSION® esiRNA, targeting human BRCA2
Sigma-Aldrich
MISSION® esiRNA, targeting human BRCA1
Sigma-Aldrich
Mirin, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-BRCA2 (Ab-1) Mouse mAb (2B), liquid, clone 2B, Calbiochem®
Roche
Cell Proliferation Kit II (XTT), liquid, pkg of 1 kit, suitable for cell analysis, suitable for tissue culture
Sigma-Aldrich
5-Chloro-2′-deoxyuridine, thymidine analog
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Hydroxyurea, 98%, powder
Sigma-Aldrich
Anti-Rad51 (Ab-1) Rabbit pAb, liquid, Calbiochem®
Sigma-Aldrich
Anti-BRCA1 (Ab-2) Mouse mAb (MS13), liquid, clone MS13, Calbiochem®