Merck
  • Mucosal Monosaccharide Transporter Expression in Newborns With Jejunoileal Atresia and Along the Adult Intestine.
Mucosal Monosaccharide Transporter Expression in Newborns With Jejunoileal Atresia and Along the Adult Intestine.
Journal of pediatric gastroenterology and nutrition (2019-07-02)
Chantal Florence Meier, Simone Mafalda Rodrigues Camargo, Schirin Hunziker, Svenja Leu, Stefan Gerhard Holland-Cunz, François Verrey, Raphael Nicolas Vuille-Dit-Bille
ABSTRACT
In newborn rodents, intestinal maturation involves delayed fructose transporter GLUT5 expression until weaning. In jejunoileal atresia (JIA), distal intestinal segments lack exposure to amniotic fluid-containing carbohydrates. We assessed in human newborns, the impact of intestinal maturation and obstruction on mucosal monosaccharide transporter expression. Samples were obtained from 10 newborns operated for small intestinal atresia and from 17 adults undergoing gastroduodenoscopy and/or ileocolonoscopy. mRNA expression of the transporters SGLT1, GLUT1, GLUT2, GLUT5, and GLUT7 was measured in neonate samples proximal and distal of the atresia as well as in adult duodenum, ileum, and colon. Protein expression and localization was assessed using immunofluorescence. Although mRNA expression of monosaccharide transporters did not significantly differ between newborn and adult samples, luminal fructose transporter GLUT5 protein was absent in 0- to 4-day-old neonates, but expressed in adults. The mRNA expression of the 5 tested monosaccharide transporters was unchanged distal from the JIA relative to proximal. Similarly, luminal sodium-dependent glucose transporter SGLT1 and basolateral GLUT2 were expressed proximal and distal to JIA as visualized by immunofluorescence staining. With the exception of glucose transporter GLUT1 that showed highest expression levels in colon, all investigated hexose transporters showed strongest expression in duodenum, lower levels in ileum and lowest in colon. Human newborns lack small intestinal fructose transporter GLUT5 protein expression and small intestinal atresia does not affect the expression of hexose transporters.
Sigma-Aldrich
Triton X-100, laboratory grade