Merck
  • Hypothalamic hamartoma simulating a suprasellar arachnoid cyst: resolution of precocious puberty following microsurgical lesion resection.
Hypothalamic hamartoma simulating a suprasellar arachnoid cyst: resolution of precocious puberty following microsurgical lesion resection.
Journal of neurosurgery. Pediatrics (2014-05-20)
Sunil Manjila, Timothy W Vogel, Yunwei Chen, Mark S Rodgers, Alan R Cohen
ABSTRACT
Hypothalamic hamartomas (HHs) are rare developmental lesions arising from the inferior hypothalamus that may cause gelastic seizures and central precocious puberty. Cystic changes in HHs are rare, usually occurring in giant lesions. The authors describe an unusual case of cystic HH masquerading as a suprasellar arachnoid cyst in an 18-month-old girl presenting with precocious puberty. Microsurgical removal of the lesion led to complete resolution of the precocious puberty on long-term follow-up. This case is the first reported HH with pathological demonstration of corticotropin-releasing hormone immunostaining in the solid tumor and glial cells in the cyst wall of the lesion. The clinical and radiological characteristics of HHs are reviewed, along with the unique surgical strategies used to manage cystic lesions in the suprasellar region.