Merck
  • 53BP1 Enforces Distinct Pre- and Post-resection Blocks on Homologous Recombination.
53BP1 Enforces Distinct Pre- and Post-resection Blocks on Homologous Recombination.
Molecular cell (2019-10-28)
Elsa Callen, Dali Zong, Wei Wu, Nancy Wong, Andre Stanlie, Momoko Ishikawa, Raphael Pavani, Lavinia C Dumitrache, Andrea K Byrum, Carlos Mendez-Dorantes, Paula Martinez, Andres Canela, Yaakov Maman, Amanda Day, Michael J Kruhlak, Maria A Blasco, Jeremy M Stark, Nima Mosammaparast, Peter J McKinnon, André Nussenzweig
ABSTRACT
53BP1 activity drives genome instability and lethality in BRCA1-deficient mice by inhibiting homologous recombination (HR). The anti-recombinogenic functions of 53BP1 require phosphorylation-dependent interactions with PTIP and RIF1/shieldin effector complexes. While RIF1/shieldin blocks 5'-3' nucleolytic processing of DNA ends, it remains unclear how PTIP antagonizes HR. Here, we show that mutation of the PTIP interaction site in 53BP1 (S25A) allows sufficient DNA2-dependent end resection to rescue the lethality of BRCA1Δ11 mice, despite increasing RIF1 "end-blocking" at DNA damage sites. However, double-mutant cells fail to complete HR, as excessive shieldin activity also inhibits RNF168-mediated loading of PALB2/RAD51. As a result, BRCA1Δ1153BP1S25A mice exhibit hallmark features of HR insufficiency, including premature aging and hypersensitivity to PARPi. Disruption of shieldin or forced targeting of PALB2 to ssDNA in BRCA1D1153BP1S25A cells restores RNF168 recruitment, RAD51 nucleofilament formation, and PARPi resistance. Our study therefore reveals a critical function of shieldin post-resection that limits the loading of RAD51.
Sigma-Aldrich
Octoclothepin maleate salt, solid
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction
Sigma-Aldrich
Doxycycline hyclate
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
ApopTag Fluorescein In Situ Apoptosis Detection Kit, The ApopTag Fluorescein In Situ Apoptosis Detection Kit detects apoptotic cells in situ by the indirect TUNEL method, utilizing an anti-digoxigenin antibody that is conjugated to a Fluorescein reporter molecule.
Sigma-Aldrich
ML216, ≥98% (HPLC)
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
Anti-Chk2 Antibody, clone 7, clone 7, Upstate®, from mouse