Merck
  • The novel F-box protein Mfb1p regulates mitochondrial connectivity and exhibits asymmetric localization in yeast.
The novel F-box protein Mfb1p regulates mitochondrial connectivity and exhibits asymmetric localization in yeast.
Molecular biology of the cell (2006-06-23)
Noriko Kondo-Okamoto, Kentaro Ohkuni, Katsumi Kitagawa, J Michael McCaffery, Janet M Shaw, Koji Okamoto
ABSTRACT
Although it is clear that mitochondrial morphogenesis is a complex process involving multiple proteins in eukaryotic cells, little is known about regulatory molecules that modulate mitochondrial network formation. Here, we report the identification of a new yeast mitochondrial morphology gene called MFB1 (YDR219C). MFB1 encodes an F-box protein family member, many of which function in Skp1-Cdc53/Cullin-F-box protein (SCF) ubiquitin ligase complexes. F-box proteins also act in non-SCF complexes whose functions are not well understood. Although cells lacking Mfb1p contain abnormally short mitochondrial tubules, Mfb1p is not essential for known pathways that determine mitochondrial morphology and dynamics. Mfb1p is peripherally associated with the mitochondrial surface. Coimmunoprecipitation assays reveal that Mfb1p interacts with Skp1p in an F-box-dependent manner. However, Mfb1p does not coimmunoprecipitate with Cdc53p. The F-box motif is not essential for Mfb1p-mediated mitochondrial network formation. These observations suggest that Mfb1p acts in a complex lacking Cdc53p required for mitochondrial morphogenesis. During budding, Mfb1p asymmetrically localizes to mother cell mitochondria. By contrast, Skp1p accumulates in the daughter cell cytoplasm. Mfb1p mother cell-specific asymmetry depends on the F-box motif, suggesting that Skp1p down-regulates Mfb1p mitochondrial association in buds. We propose that Mfb1p operates in a novel pathway regulating mitochondrial tubular connectivity.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)