Merck
  • Synaptogenesis Stimulates a Proteasome-Mediated Ribosome Reduction in Axons.
Synaptogenesis Stimulates a Proteasome-Mediated Ribosome Reduction in Axons.
Cell reports (2019-07-25)
Rui O Costa, Helena Martins, Luís F Martins, Andrzej W Cwetsch, Miranda Mele, Joana R Pedro, Diogo Tomé, Noo Li Jeon, Laura Cancedda, Samie R Jaffrey, Ramiro D Almeida
ABSTRACT
Ribosomes and a subset of cellular mRNAs are trafficked into axons of developing neurons. The axonal localization of translational machinery allows new proteins to be rapidly and locally synthesized during axonal growth and pathfinding. However, in mature neurons, axonal ribosomes are significantly reduced or even absent. The mechanism that elicits this removal is currently unknown. Here, we demonstrate that synapse formation is the trigger for ribosome reduction in mature axons. In vivo analysis shows that axonal ribosome levels decrease in rat brain at a developmental stage coincident with synapse formation. Next, we observe in vitro that different synaptogenic inducers trigger an overall decrease of ribosomal proteins and rRNA in the axons of spinal motor neurons. We further observe that this process is dependent on the ubiquitin-proteasome system but not on autophagy. Together, these data identify synaptogenesis as the long missing biological trigger that leads to ribosome disappearance during axonal maturation.
Sigma-Aldrich
Leupeptin, microbial, ≥90% (HPLC)
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Anti-Neurofilament M Antibody, Chemicon®, from chicken
Sigma-Aldrich
Anti-Synapsin I Antibody, serum, Chemicon®