Merck
  • Inactivation of the glutathione peroxidase GPx4 by the ferroptosis-inducing molecule RSL3 requires the adaptor protein 14-3-3ε.
Inactivation of the glutathione peroxidase GPx4 by the ferroptosis-inducing molecule RSL3 requires the adaptor protein 14-3-3ε.
FEBS letters (2019-10-04)
Ana-Marija Vučković, Valentina Bosello Travain, Luciana Bordin, Giorgio Cozza, Giovanni Miotto, Monica Rossetto, Stefano Toppo, Rina Venerando, Mattia Zaccarin, Matilde Maiorino, Fulvio Ursini, Antonella Roveri
ABSTRACT
Ras-selective lethal small molecule 3 (RSL3), a drug candidate prototype for cancer chemotherapy, triggers ferroptosis by inactivating the glutathione peroxidase glutathione peroxidase 4 (GPx4). Here, we report the purification of the protein indispensable for GPx4 inactivation by RSL3. Mass spectrometric analysis identified 14-3-3 isoforms as candidates, and recombinant human 14-3-3ε confirms the identification. The function of 14-3-3ε is redox-regulated. Moreover, overexpression or silencing of the gene coding for 14-3-3ε consistently controls the inactivation of GPx4 by RSL3. The interaction of GPx4 with a redox-regulated adaptor protein operating in cell signaling further contributes to frame it within redox-regulated pathways of cell survival and death and opens new therapeutic perspectives.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
MISSION® esiRNA, targeting human YWHAE
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule), F(ab′)2 fragment−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution