Merck
  • Low-Level Saturated Fatty Acid Palmitate Benefits Liver Cells by Boosting Mitochondrial Metabolism via CDK1-SIRT3-CPT2 Cascade.
Low-Level Saturated Fatty Acid Palmitate Benefits Liver Cells by Boosting Mitochondrial Metabolism via CDK1-SIRT3-CPT2 Cascade.
Developmental cell (2019-12-24)
Lin Liu, Bowen Xie, Ming Fan, Demet Candas-Green, Joy X Jiang, Ryan Wei, Yinsheng Wang, Hong-Wu Chen, Yiyang Hu, Jian Jian Li
ABSTRACT
Saturated fatty acids (SFAs) (the "bad" fat), especially palmitate (PA), in the human diet are blamed for potential health risks such as obesity and cancer because of SFA-induced lipotoxicity. However, epidemiological results demonstrate a latent benefit of SFAs, and it remains elusive whether a certain low level of SFAs is physiologically essential for maintaining cell metabolic hemostasis. Here, we demonstrate that although high-level PA (HPA) indeed induces lipotoxic effects in liver cells, low-level PA (LPA) increases mitochondrial functions and alleviates the injuries induced by HPA or hepatoxic agent carbon tetrachloride (CCl4). LPA treatment in mice enhanced liver mitochondrial activity and reduced CCl4 hepatotoxicity with improved blood levels of aspartate aminotransferase (AST), alanine transaminase (ALT), and mitochondrial aspartate transaminase (m-AST). LPA-mediated mitochondrial homeostasis is regulated by CDK1-mediated SIRT3 phosphorylation, which in turn deacetylates and dimerizes CPT2 to enhance fatty acid oxidation. Thus, an advantageous effect is suggested by the consumption of LPA that augments mitochondrial metabolic homeostasis via CDK1-SIRT3-CPT2 cascade.
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)