Merck
  • Loss of PGC-1α in RPE induces mesenchymal transition and promotes retinal degeneration.
Loss of PGC-1α in RPE induces mesenchymal transition and promotes retinal degeneration.
Life science alliance (2019-05-19)
Mariana Aparecida Brunini Rosales, Daisy Y Shu, Jared Iacovelli, Magali Saint-Geniez
ABSTRACT
The retinal pigment epithelium (RPE) supports visual processing and photoreceptor homeostasis via energetically demanding cellular functions. Here, we describe the consequences of repressing peroxisome proliferator-activated receptor γ coactivator-1 α (PGC-1α), a master regulator of mitochondrial function and biogenesis, on RPE epithelial integrity. The sustained silencing of PGC-1α in differentiating human RPE cells affected mitochondria/autophagy function, redox state, and impaired energy sensor activity ultimately inducing epithelial to mesenchymal transition (EMT). Adult conditional knockout of PGC-1 coactivators in mice resulted in rapid RPE dysfunction and transdifferentiation associated with severe photoreceptor degeneration. RPE anomalies were characteristic of autophagic defect and mesenchymal transition comparable with the ones observed in age-related macular degeneration. These findings demonstrate that PGC-1α is required to maintain the functional and phenotypic status of RPE by supporting the cells' oxidative metabolism and autophagy-mediated repression of EMT.
Sigma-Aldrich
Anti-PGC-1α Mouse mAb (4C1.3), liquid, clone 4C1.3, Calbiochem®
Sigma-Aldrich
MBP, Dephosphorylated
Sigma-Aldrich
Anti-Apolipoprotein E Antibody, serum, Chemicon®