Merck
Integrin expression regulates neuroblastoma attachment and migration.
Neoplasia (New York, N.Y.) (2004-07-17)
Amy Meyer, Cynthia M van Golen, Bhumsoo Kim, Kenneth L van Golen, Eva L Feldman
ABSTRACT
Neuroblastoma (NBL) is the most common malignant disease of infancy, and children with bone metastasis have a mortality rate greater than 90%. Two major classes of proteins, integrins and growth factors, regulate the metastatic process. We have previously shown that tumorigenic NBL cells express higher levels of the type I insulin-like growth factor receptor (IGF-IR) and that beta1 integrin expression is inversely proportional to tumorigenic potential in NBL. In the current study, we analyze the effect of beta1 integrin and IGF-IR on NBL cell attachment and migration. Nontumorigenic S-cells express high levels of beta1 integrin, whereas tumorigenic N-cells express little beta1 integrin. Alterations in beta1 integrin are due to regulation at the protein level, as translation is decreased in N-type cells. Moreover, inhibition of protein synthesis shows that beta1 integrin is degraded more slowly in S-type cells (SHEP) than in N-type cells (SH-SY5Y and IMR32). Inhibition of alpha5beta1 integrin prevents SHEP (but not SH-SY5Y or IMR32) cell attachment to fibronectin and increases SHEP cell migration. Increases in IGF-IR decrease beta1 integrin expression, and enhance SHEP cell migration, potentially through increased expression of alphavbeta3. These data suggest that specific classes of integrins in concert with IGF-IR regulate NBL attachment and migration.
Sigma-Aldrich
Anti-Integrin α5 Antibody, clone P1D6, azide free, clone P1D6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α1 Antibody, clone FB12, azide free, clone FB12, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α3 Antibody, clone P1B5, azide free, clone P1B5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, FITC conjugated, clone LM609, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α2 Antibody, clone P1E6, azide free, clone P1E6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α4 Antibody, clone P1H4, azide free, clone P1H4, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone HB1.1, clone HB1.1, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin αV Antibody, clone P3G8, azide free, clone P3G8, Chemicon®, from mouse