MilliporeSigma
  • The P2Y2 nucleotide receptor mediates UTP-induced vascular cell adhesion molecule-1 expression in coronary artery endothelial cells.
The P2Y2 nucleotide receptor mediates UTP-induced vascular cell adhesion molecule-1 expression in coronary artery endothelial cells.
The Journal of biological chemistry (2003-04-26)
Cheikh I Seye, Ningpu Yu, Renu Jain, Qiongman Kong, Tess Minor, Jessica Newton, Laurie Erb, Fernando A González, Gary A Weisman
ABSTRACT
P2Y2 receptor up-regulation and activation induces intimal hyperplasia and monocyte/macrophage infiltration in the collared rabbit carotid artery model of vascular injury, suggesting a potential role for P2Y2 receptors in monocyte recruitment by vascular endothelium. In this study, we addressed the hypothesis that activation of P2Y2 receptors by extracellular nucleotides modulates the expression of adhesion molecules on vascular endothelial cells that are important for monocyte recruitment. Results indicated that the equipotent P2Y2 receptor agonists UTP or ATP (1-100 microm) stimulated the expression of vascular cell adhesion molecule-1 (VCAM-1) in human coronary artery endothelial cells (HCAEC) in a time- and dose-dependent manner. P2Y2 antisense oligonucleotides inhibited VCAM-1 expression induced by UTP but not by tumor necrosis factor-alpha. Furthermore, UTP induced VCAM-1 expression in human 1321N1 astrocytoma cell transfectants expressing the recombinant P2Y2 receptor, whereas vector-transfected control cells did not respond to UTP. The effect of UTP on VCAM-1 expression in HCAEC was prevented by depletion of intracellular calcium stores with thapsigargin or by inhibition of p38 mitogen-activated protein kinase or Rho kinase, but was not affected by inhibitors of the mitogen-activated protein/extracellular signal-regulated kinase pathway (i.e. MEK1/2). Consistent with a role for VCAM-1 in the recruitment of monocytes, UTP or ATP increased the adherence of monocytic U937 cells to HCAEC, an effect that was inhibited by anti-VCAM-1 antibodies. These findings suggest a novel role for the P2Y2 receptor in the p38- and Rho kinase-dependent expression of VCAM-1 that mediates the recruitment of monocytes by vascular endothelium associated with the development of atherosclerosis.
Sigma-Aldrich
Anti-ICAM-1 Antibody, clone P2A4, clone P2A4, Chemicon®, from mouse