Merck
  • GPR56/ADGRG1 regulates development and maintenance of peripheral myelin.
GPR56/ADGRG1 regulates development and maintenance of peripheral myelin.
The Journal of experimental medicine (2018-01-26)
Sarah D Ackerman, Rong Luo, Yannick Poitelon, Amit Mogha, Breanne L Harty, Mitchell D'Rozario, Nicholas E Sanchez, Asvin K K Lakkaraju, Paul Gamble, Jun Li, Jun Qu, Matthew R MacEwan, Wilson Zachary Ray, Adriano Aguzzi, M Laura Feltri, Xianhua Piao, Kelly R Monk
ABSTRACT
Myelin is a multilamellar sheath generated by specialized glia called Schwann cells (SCs) in the peripheral nervous system (PNS), which serves to protect and insulate axons for rapid neuronal signaling. In zebrafish and rodent models, we identify GPR56/ADGRG1 as a conserved regulator of PNS development and health. We demonstrate that, during SC development, GPR56-dependent RhoA signaling promotes timely radial sorting of axons. In the mature PNS, GPR56 is localized to distinct SC cytoplasmic domains, is required to establish proper myelin thickness, and facilitates organization of the myelin sheath. Furthermore, we define plectin-a scaffolding protein previously linked to SC domain organization, myelin maintenance, and a series of disorders termed "plectinopathies"-as a novel interacting partner of GPR56. Finally, we show that Gpr56 mutants develop progressive neuropathy-like symptoms, suggesting an underlying mechanism for peripheral defects in some human patients with GPR56 mutations. In sum, we define Gpr56 as a new regulator in the development and maintenance of peripheral myelin.
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Human Collagen Type III, 1 mg/mL, solution, Bioreagent, suitable for cell culture
Sigma-Aldrich
Anti-Plectin Antibody, Clone 10F6, clone 10F6, Chemicon®, from mouse