Merck
  • Specific contributions of N-methyl-D-aspartate receptors in the dorsal striatum to cognitive flexibility.
Specific contributions of N-methyl-D-aspartate receptors in the dorsal striatum to cognitive flexibility.
Neuroscience (2014-12-03)
M Darvas, R D Palmiter
ABSTRACT
Behavioral flexibility is known to be mediated by corticostriatal systems and to involve several major neurotransmitter signaling pathways. The current study investigated the effects of inactivation of glutamatergic N-methyl-D-aspartate-(NMDA) receptor signaling in the dorsal striatum on behavioral flexibility in mice. NMDA-receptor inactivation was achieved by virus-mediated inactivation of the Grin1 gene, which encodes the essential NR1 subunit of NMDA receptors. To assess behavioral flexibility, we used a water U-maze paradigm in which mice had to shift from an initially acquired rule to a new rule (strategy shifting) or had to reverse an initially learned rule (reversal learning). Inactivation of NMDA-receptors in all neurons of the dorsal striatum did not affect learning of the initial rule or reversal learning, but impaired shifting from one strategy to another. Strategy shifting was also compromised when NMDA-receptors were inactivated only in dynorphin-expressing neurons in the dorsal striatum, which represent the direct pathway. These data suggest that NMDA-receptor-mediated synaptic plasticity in the dorsal striatum contributes to strategy shifting and that striatal projection neurons of the direct pathway are particularly relevant for this process.
Sigma-Aldrich
Anti-NMDAR1 Antibody, clone 54.1, clone 54.1, Chemicon®, from mouse