Merck
  • Hexacosenoyl-CoA is the most abundant very long-chain acyl-CoA in ATP binding cassette transporter D1-deficient cells.
Hexacosenoyl-CoA is the most abundant very long-chain acyl-CoA in ATP binding cassette transporter D1-deficient cells.
Journal of lipid research (2020-02-23)
Kotaro Hama, Yuko Fujiwara, Shigeo Takashima, Yasuhiro Hayashi, Atsushi Yamashita, Nobuyuki Shimozawa, Kazuaki Yokoyama
ABSTRACT
X-linked adrenoleukodystrophy (X-ALD) is an inherited disorder caused by deleterious mutations in the ABCD1 gene. The ABCD1 protein transports very long-chain FAs (VLCFAs) from the cytosol into the peroxisome where the VLCFAs are degraded through β-oxidation. ABCD1 dysfunction leads to VLCFA accumulation in individuals with X-ALD. FAs are activated by esterification to CoA before metabolic utilization. However, the intracellular pools and metabolic profiles of individual acyl-CoA esters have not been fully analyzed. In this study, we profiled the acyl-CoA species in fibroblasts from X-ALD patients and in ABCD1-deficient HeLa cells. We found that hexacosenoyl (26:1)-CoA, but not hexacosanoyl (26:0)-CoA, was the most abundantly concentrated among the VLCFA-CoA species in these cells. We also show that 26:1-CoA is mainly synthesized from oleoyl-CoA, and the metabolic turnover rate of 26:1-CoA was almost identical to that of oleoyl-CoA in both WT and ABCD1-deficient HeLa cells. The findings of our study provide precise quantitative and metabolic information of each acyl-CoA species in living cells. Our results suggest that VLCFA is endogenously synthesized as VLCFA-CoA through a FA elongation pathway and is then efficiently converted to other metabolites, such as phospholipids, in the absence of ABCD1.
Product Description
Avanti
16:0-d31-18:1 PE, 1-palmitoyl-d31-2-oleoyl-sn-glycero-3-phosphoethanolamine, powder
Avanti
16:0-d31-18:1 PE, 1-palmitoyl-d31-2-oleoyl-sn-glycero-3-phosphoethanolamine, chloroform
Avanti
16:0-d31-18:1 PC, 1-palmitoyl-d31-2-oleoyl-sn-glycero-3-phosphocholine, chloroform
Avanti
16:0-d31 SM, Avanti Research - A Croda Brand
Avanti
26:0 Coenzyme A, Avanti Research - A Croda Brand 870726P, powder
Avanti
16:0-d31-18:1 PC, 1-palmitoyl-d31-2-oleoyl-sn-glycero-3-phosphocholine, powder