Merck
  • A novel multicommuted fluorimetric optosensor for determination of resveratrol in beer.
A novel multicommuted fluorimetric optosensor for determination of resveratrol in beer.
Talanta (2010-12-15)
Lucía Molina-García, Antonio Ruiz-Medina, Maria Luisa Fernández-de Córdova
ABSTRACT
Resveratrol (3,5,4'-trihydroxystilbene) is a phytoalexin that plays a central role in the human diet because of its antioxidant, anticarcinogenic and antimutagenic properties. This paper shows the development of a multicommuted optosensing device for the determination of resveratrol in beer. The method is based on the measurement of the fluorescence (277/382 nm, λ(ex)/λ(em)) of the photoproduct on-line generated by UV-irradiation of resveratrol. The fluorescent photoproduct is monitored once it is retained on a solid support (Sephadex QAE A-25) in the detection area, which improves both sensitivity and selectivity. The sample was delipidated with toluene and cyclohexane and resveratrol was extracted by solid-phase extraction (SPE) on C(18) cartridges, using methanol as eluent. This pre-treatment allowed recovering about an 82% resveratrol and removing its 3-O-β-d-glycoside (piceid) and other interfering substances present in beer. The method provides a detection limit (DL) of 1.0 ng mL(-1) and a linear dynamic range (LDR) of 3.3-100 ng mL(-1). It was satisfactorily applied to the determination of resveratrol in top- and bottom-fermented beers by standard addition calibration. Resveratrol concentrations in the analysed samples varied from 4.1 to 14.1 ng mL(-1). This is the first proposed spectroscopic method for determination of resveratrol in beer.
Sigma-Aldrich
QAE Sephadex® A-25 chloride form
Sigma-Aldrich
QAE Sephadex® A-50 chloride form