Merck
Anti-inflammatory and antifibrotic effects of methyl palmitate.
Toxicology and applied pharmacology (2011-05-18)
Ebtehal El-Demerdash
ABSTRACT
Methyl palmitate (MP) has been shown earlier to inhibit Kupffer cells and rat peritoneal macrophages. To evaluate the potential of MP to inhibit the activation of other macrophages, RAW cells (macrophages of alveolar origin) were treated with varying concentrations of MP (0.25, 0.5, 1mM). Assessment of cytotoxicity using MTT assay revealed that 0.25 and 0.5mM are not toxic to RAW cells. MP was able to inhibit the phagocytic function of RAW cells. Treatment of cells with MP 24hours prior to LPS stimulation significantly decreased nitric oxide release and altered the pattern of cytokines release; there was a significant decrease in TNF-α and a significant increase in IL-10 compared to the controls. However, there is a non-significant change in IL-6 level. Furthermore, phosphorylation of inhibitory kappa B (IκBα) protein was significantly decreased in RAW cells treated with 0.5mM MP after LPS stimulation. Based upon the in-vitro results, it was examined whether MP treatment will be effective in preventing bleomycin-induced lung inflammation and fibrosis in-vivo. Bleomycin given by itself caused destruction of the lung architecture characterized by pulmonary fibrosis with collapse of air alveoli and emphysematous. Bleomycin induced a significant increase in hydroxyproline level and activated NF-κB, p65 expression in the lung. MP co-treatment significantly ameliorated bleomycin effects. These results suggest that MP has a potential of inhibiting macrophages in general. The present study demonstrated for the first time that MP has anti-inflammatory and antifibrotic effect that could be through NF-kB inhibition. Thus MP like molecule could be a promising anti-inflammatory and antifibrotic drug.
Sigma-Aldrich
Methyl palmitate, ≥99% (capillary GC)
Supelco
Methyl palmitate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Methyl palmitate, analytical standard
Supelco
Methyl Palmitate, pharmaceutical secondary standard, certified reference material
Sigma-Aldrich
Methyl palmitate, ≥97%
USP
Methyl palmitate, United States Pharmacopeia (USP) Reference Standard