Merck
  • Peroxisomes as Modulators of Cellular Protein Thiol Oxidation: A New Model System.
Peroxisomes as Modulators of Cellular Protein Thiol Oxidation: A New Model System.
Antioxidants & redox signaling (2017-06-09)
Celien Lismont, Marcus Nordgren, Chantal Brees, Bernard Knoops, Paul P Van Veldhoven, Marc Fransen
ABSTRACT
Peroxisomes are ubiquitous, single-membrane-bounded organelles that contain considerable amounts of enzymes involved in the production or breakdown of hydrogen peroxide (H2O2), a key signaling molecule in multiple biological processes and disease states. Despite this, the role of this organelle in cross-compartmental H2O2 signaling remains largely unclear, mainly because of the difficulty to modulate peroxisomal H2O2 production in a selective manner. This study aimed at establishing and validating a cellular model suitable to decipher the complex signaling processes associated with peroxisomal H2O2 release. Here, we report the development of a human cell line that can be used to selectively generate H2O2 inside peroxisomes in a time- and dose-controlled manner. In addition, we provide evidence that peroxisome-derived H2O2 can oxidize redox-sensitive cysteine residues in multiple proteins within (e.g., peroxiredoxin-5 [PRDX5]) and outside (e.g., nuclear factor kappa B subunit 1 [NFKB1] and subunit RELA proto-oncogene [RELA], phosphatase and tensin homolog [PTEN], forkhead box O3 [FOXO3], and peroxin 5 [PEX5]) the peroxisomal compartment. Furthermore, we show that the extent of protein oxidation depends on the subcellular location of the target protein and is inversely correlated to catalase activity and cellular glutathione content. Finally, we demonstrate that excessive H2O2 production inside peroxisomes does not induce their selective degradation, at least not under the conditions examined. This study describes for the first time a powerful model system that can be used to examine the role of peroxisome-derived H2O2 in redox-regulated (patho)physiological processes, a research area in need of further investigation and innovative approaches. Our results provide unambiguous evidence that peroxisomes can serve as regulatory hubs in thiol-based signaling networks.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
L-Buthionine-sulfoximine, ≥97% (TLC)
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution