Merck
  • Starvation induces rapid degradation of selective autophagy receptors by endosomal microautophagy.
Starvation induces rapid degradation of selective autophagy receptors by endosomal microautophagy.
The Journal of cell biology (2018-07-19)
Jakob Mejlvang, Hallvard Olsvik, Steingrim Svenning, Jack-Ansgar Bruun, Yakubu Princely Abudu, Kenneth Bowitz Larsen, Andreas Brech, Tom E Hansen, Hanne Brenne, Terkel Hansen, Harald Stenmark, Terje Johansen
ABSTRACT
It is not clear to what extent starvation-induced autophagy affects the proteome on a global scale and whether it is selective. In this study, we report based on quantitative proteomics that cells during the first 4 h of acute starvation elicit lysosomal degradation of up to 2-3% of the proteome. The most significant changes are caused by an immediate autophagic response elicited by shortage of amino acids but executed independently of mechanistic target of rapamycin and macroautophagy. Intriguingly, the autophagy receptors p62/SQSTM1, NBR1, TAX1BP1, NDP52, and NCOA4 are among the most efficiently degraded substrates. Already 1 h after induction of starvation, they are rapidly degraded by a process that selectively delivers autophagy receptors to vesicles inside late endosomes/multivesicular bodies depending on the endosomal sorting complex required for transport III (ESCRT-III). Our data support a model in which amino acid deprivation elicits endocytosis of specific membrane receptors, induction of macroautophagy, and rapid degradation of autophagy receptors by endosomal microautophagy.
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Hanks′ Balanced Salt solution, Modified, with sodium bicarbonate, without phenol red, liquid, sterile-filtered, suitable for cell culture