Merck
  • Promotion of growth factor signaling as a critical function of β-catenin during HCC progression.
Promotion of growth factor signaling as a critical function of β-catenin during HCC progression.
Nature communications (2019-04-25)
Eunsun Kim, Amanda Lisby, Connie Ma, Nathanael Lo, Ursula Ehmer, Katharina E Hayer, Emma E Furth, Patrick Viatour
ABSTRACT
Hepatocellular carcinoma (HCC) is the second leading cause of cancer-related deaths worldwide. β-catenin is widely thought to be a major oncogene in HCC based on the frequency of mutations associated with aberrant Wnt signaling in HCC patients. Challenging this model, our data reveal that β-catenin nuclear accumulation is restricted to the late stage of the disease. Until then, β-catenin is primarily located at the plasma membrane in complex with multiple cadherin family members where it drives tumor cell survival by enhancing the signaling of growth factor receptors such as EGFR. Therefore, our study reveals the evolving nature of β-catenin in HCC to establish it as a compound tumor promoter during the progression of the disease.
Sigma-Aldrich
Anti-Axin-1 Antibody, clone A5, clone A5, from mouse
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)