Merck
  • Using glycyrrhizic acid to target sumoylation processes during Epstein-Barr virus latency.
Using glycyrrhizic acid to target sumoylation processes during Epstein-Barr virus latency.
PloS one (2019-05-28)
Gretchen L Bentz, Angela J Lowrey, Dustin C Horne, Vy Nguyen, Austin R Satterfield, Tabithia D Ross, Abigail E Harrod, Olga N Uchakina, Robert J McKallip
ABSTRACT
Cellular sumoylation processes are proposed targets for anti-viral and anti-cancer therapies. We reported that Epstein-Barr virus (EBV) latent membrane protein 1 (LMP1) dysregulates cellular sumoylation processes, contributing to its oncogenic potential in EBV-associated malignancies. Ginkgolic acid and anacardic acid, known inhibitors of sumoylation, inhibit LMP1-induced protein sumoylation; however, both drugs have adverse effects in hosts. Here we test the effects of glycyrrhizic acid, a medicinal botanical extract with anti-inflammatory, anti-carcinogenic, and anti-viral properties, on cellular sumoylation processes. While glycyrrhizic acid is known to inhibit EBV penetration, its affect on cellular sumoylation processes remains to be documented. We hypothesized that glycyrrhizic acid inhibits cellular sumoylation processes and may be a viable treatment for Epstein-Barr virus-associated malignancies. Results showed that glycyrrhizic acid inhibited sumoylation processes (without affecting ubiquitination processes), limited cell growth, and induced apoptosis in multiple cell lines. Similar to ginkgolic acid; glycyrrhizic acid targeted the first step of the sumoylation process and resulted in low levels of spontaneous EBV reactivation. Glycyrrhizic acid did not affect induced reactivation of the virus, but the presence of the extract did reduce the ability of the produced virus to infect additional cells. Therefore, we propose that glycyrrhizic acid may be a potential therapeutic drug to augment the treatment of EBV-associated lymphoid malignancies.