Merck
  • Vaccination establishes clonal relatives of germinal center T cells in the blood of humans.
Vaccination establishes clonal relatives of germinal center T cells in the blood of humans.
The Journal of experimental medicine (2017-06-24)
Antje Heit, Frank Schmitz, Sarah Gerdts, Britta Flach, Miranda S Moore, Jonathan A Perkins, Harlan S Robins, Alan Aderem, Paul Spearman, Georgia D Tomaras, Stephen C De Rosa, M Juliana McElrath
ABSTRACT
Germinal center T follicular helper cells (GCTfh) in lymphatic tissue are critical for B cell differentiation and protective antibody induction, but whether GCTfh establish clonal derivatives as circulating memory T cells is less understood. Here, we used markers expressed on GCTfh, CXCR5, PD1, and ICOS, to identify potential circulating CXCR5+CD4+ Tfh-like cells (cTfh) in humans, and investigated their functional phenotypes, diversity, and ontogeny in paired donor blood and tonsils, and in blood after vaccination. Based on T cell receptor repertoire analysis, we found that PD-1-expressing cTfh and tonsillar GCTfh cells were clonally related. Furthermore, an activated, antigen-specific PD1+ICOS+ cTfh subset clonally expanded after booster immunization whose frequencies correlated with vaccine-specific serum IgG; these phenotypically resembled GCTfh, and were clonally related to a resting PD1+ICOS- CD4+ memory T cell subset. Thus, we postulate that vaccination establishes clonal relatives of GCTfh within the circulating memory CD4+CXCR5+PD1+ T cell pool that expand upon reencounter of their cognate antigen.
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)