Merck
  • ATP-binding cassette transporter expression in human placenta as a function of pregnancy condition.
ATP-binding cassette transporter expression in human placenta as a function of pregnancy condition.
Drug metabolism and disposition: the biological fate of chemicals (2011-03-25)
Cifford W Mason, Irina A Buhimschi, Catalin S Buhimschi, Yafeng Dong, Carl P Weiner, Peter W Swaan
ABSTRACT
Fetal drug exposure is determined by the type and concentration of placental transporters, and their regulation is central to the development of new treatments and delivery strategies for pregnant women and their fetuses. We tested the expression of several clinically important transporters in the human placenta associated with various pregnancy conditions (i.e., labor, preeclampsia, and preterm labor-inflammation). Placentas were obtained from five groups of women at the time of primary cesarean section: 1) term no labor; 2) term labor; 3) preterm no labor (delivered for severe preeclampsia); 4) preterm labor without inflammation (PTLNI); and 5) preterm labor with inflammation (PTLI). Samples were analyzed by Western blot and immunohistochemistry to identify changes in protein expression. Relative mRNA expression was determined by quantitative real-time polymerase chain reaction. A functional genomic approach was used to identify placental gene expression and elucidate molecular events that underlie the given condition. Placental expression of ATP-binding cassette transporters from women in labor and women with preeclampsia was unaltered. Multidrug resistance protein 1 (MDR1) and breast cancer resistance protein (BCRP) and mRNA expression increased in placentas of women with preterm labor with inflammation. Molecular pathways of genes up-regulated in PTLI samples included cytokine-cytokine receptor interactions and inflammatory response compared with those in the PTLNI group. The mRNA expression of MDR1 and BCRP was correlated with that of interleukin-8, which also increased significantly in PTLI samples. These data suggest that the transfer of drugs across the placenta may be altered in preterm pregnancy conditions associated with inflammation through changes in MDR1 and BCRP.
Sigma-Aldrich
Anti-MRP2 Antibody, CT, clone M2 III-6, culture supernatant, clone M2 III-6, Chemicon®
Sigma-Aldrich
Anti-MDR3 Antibody, clone P3 II-26, culture supernatant, clone P3 II-26, Chemicon®