Merck
  • Human organotypic brain slice culture: a novel framework for environmental research in neuro-oncology.
Human organotypic brain slice culture: a novel framework for environmental research in neuro-oncology.
Life science alliance (2019-06-30)
Vidhya M Ravi, Kevin Joseph, Julian Wurm, Simon Behringer, Nicklas Garrelfs, Paolo d'Errico, Yashar Naseri, Pamela Franco, Melanie Meyer-Luehmann, Roman Sankowski, Mukesch Johannes Shah, Irina Mader, Daniel Delev, Marie Follo, Jürgen Beck, Oliver Schnell, Ulrich G Hofmann, Dieter Henrik Heiland
ABSTRACT
When it comes to the human brain, models that closely mimic in vivo conditions are lacking. Living neuronal tissue is the closest representation of the in vivo human brain outside of a living person. Here, we present a method that can be used to maintain therapeutically resected healthy neuronal tissue for prolonged periods without any discernible changes in tissue vitality, evidenced by immunohistochemistry, genetic expression, and electrophysiology. This method was then used to assess glioblastoma (GBM) progression in its natural environment by microinjection of patient-derived tumor cells into cultured sections. The result closely resembles the pattern of de novo tumor growth and invasion, drug therapy response, and cytokine environment. Reactive transformation of astrocytes, as an example of the cellular nonmalignant tumor environment, can be accurately simulated with transcriptional differences similar to those of astrocytes isolated from acute GBM specimens. In a nutshell, we present a simple method to study GBM in its physiological environment, from which valuable insights can be gained. This technique can lead to further advancements in neuroscience, neuro-oncology, and pharmacotherapy.
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Methyl-D-glucamine, 99.0-100.5% (titration)
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse