Merck
  • Nucleocytoplasmic shuttling of the zinc finger protein EZI Is mediated by importin-7-dependent nuclear import and CRM1-independent export mechanisms.
Nucleocytoplasmic shuttling of the zinc finger protein EZI Is mediated by importin-7-dependent nuclear import and CRM1-independent export mechanisms.
The Journal of biological chemistry (2007-09-13)
Eiko Saijou, Tohru Itoh, Kyung-Woon Kim, Shun-ichiro Iemura, Tohru Natsume, Atsushi Miyajima
ABSTRACT
Nucleocytoplasmic translocation constitutes a foundation for nuclear proteins to exert their proper functions and hence for various biological reactions to occur normally in eukaryotic cells. We reported previously that EZI/Zfp467, a 12 zinc finger motif-containing protein, localizes predominantly in the nucleus, yet the underlying mechanism still remains elusive. Here we constructed a series of mutant forms of EZI and examined their subcellular localization. The results delineated a non-canonical nuclear localization signal in the region covering the 9th to the 12th zinc fingers, which was necessary for nuclear accumulation of EZI as well as sufficient to confer nuclear localizing ability to a heterologous protein. We also found that the N-terminal domain of EZI is necessary for its nuclear export, the process of which was not sensitive to the CRM1 inhibitor leptomycin B. An interaction proteomics approach and the following co-immunoprecipitation experiments identified the nuclear import receptor importin-7 as a molecule that associated with EZI and, importantly, short interfering RNA-mediated knockdown of importin-7 expression completely abrogated nuclear accumulation of EZI. Taken together, these results identify EZI as a novel cargo protein for importin-7 and demonstrate a nucleocytoplasmic shuttling mechanism that is mediated by importin-7-dependent nuclear localization and CRM1-independent nuclear export.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)