Merck
Identification of a novel fusion gene HMGA2-EGFR in glioblastoma.
International journal of cancer (2017-12-02)
Akiyoshi Komuro, Erna Raja, Caname Iwata, Manabu Soda, Kazunobu Isogaya, Keiko Yuki, Yasushi Ino, Masato Morikawa, Tomoki Todo, Hiroyuki Aburatani, Hiromichi Suzuki, Melissa Ranjit, Atsushi Natsume, Akitake Mukasa, Nobuhito Saito, Hitoshi Okada, Hiroyuki Mano, Kohei Miyazono, Daizo Koinuma
ABSTRACT
Glioblastoma is one of the most malignant forms of cancer, for which no effective targeted therapy has been found. Although The Cancer Genome Atlas has provided a list of fusion genes in glioblastoma, their role in progression of glioblastoma remains largely unknown. To search for novel fusion genes, we obtained RNA-seq data from TGS-01 human glioma-initiating cells, and identified a novel fusion gene (HMGA2-EGFR), encoding a protein comprising the N-terminal region of the high-mobility group AT-hook protein 2 (HMGA2) fused to the C-terminal region of epidermal growth factor receptor (EGFR), which retained the transmembrane and kinase domains of the EGFR. This fusion gene product showed transforming potential and a high tumor-forming capacity in cell culture and in vivo. Mechanistically, HMGA2-EGFR constitutively induced a higher level of phosphorylated STAT5B than EGFRvIII, an in-frame exon deletion product of the EGFR gene that is commonly found in primary glioblastoma. Forced expression of HMGA2-EGFR enhanced orthotopic tumor formation of the U87MG human glioma cell line. Furthermore, the EGFR kinase inhibitor erlotinib blocked sphere formation of TGS-01 cells in culture and inhibited tumor formation in vivo. These findings suggest that, in addition to gene amplification and in-frame exon deletion, EGFR signaling can also be activated by gene fusion, suggesting a possible avenue for treatment of glioblastoma.
Product Description
Millipore
JAK Inhibitor I, JAK Inhibitor I, CAS 457081-03-7, is a potent, reversible, cell-permeable, and ATP-competitive inhibitor of JAK 1 (IC50 = 15 nM), JAK2 (IC50 = 1 nM), JAK3 (Ki = 5 nM) and Tyk2 (IC50 = 1 nM).
Sigma-Aldrich
STAT3 Inhibitor III, WP1066, STAT3 Inhibitor III, WP1066, CAS 857064-38-1, is a cell-permeable AG490 tyrphostin analog that acts as a STAT3 pathway inhibitor. A potent anti-tumor agent.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
JAK2 Inhibitor III, SD-1029, The JAK2 Inhibitor III, SD-1029, also referenced under CAS 118372-34-2, controls the biological activity of JAK2.
Sigma-Aldrich
Anti-EGFR (cytoplasmic domain) Antibody, clone 8G6.2, clone 8G6.2, from mouse