Merck
  • Suppression of p16 Induces mTORC1-Mediated Nucleotide Metabolic Reprogramming.
Suppression of p16 Induces mTORC1-Mediated Nucleotide Metabolic Reprogramming.
Cell reports (2019-08-23)
Raquel Buj, Chi-Wei Chen, Erika S Dahl, Kelly E Leon, Rostislav Kuskovsky, Natella Maglakelidze, Maithili Navaratnarajah, Gao Zhang, Mary T Doan, Helen Jiang, Michael Zaleski, Lydia Kutzler, Holly Lacko, Yiling Lu, Gordon B Mills, Raghavendra Gowda, Gavin P Robertson, Joshua I Warrick, Meenhard Herlyn, Yuka Imamura, Scot R Kimball, David J DeGraff, Nathaniel W Snyder, Katherine M Aird
ABSTRACT
Reprogrammed metabolism and cell cycle dysregulation are two cancer hallmarks. p16 is a cell cycle inhibitor and tumor suppressor that is upregulated during oncogene-induced senescence (OIS). Loss of p16 allows for uninhibited cell cycle progression, bypass of OIS, and tumorigenesis. Whether p16 loss affects pro-tumorigenic metabolism is unclear. We report that suppression of p16 plays a central role in reprogramming metabolism by increasing nucleotide synthesis. This occurs by activation of mTORC1 signaling, which directly mediates increased translation of the mRNA encoding ribose-5-phosphate isomerase A (RPIA), a pentose phosphate pathway enzyme. p16 loss correlates with activation of the mTORC1-RPIA axis in multiple cancer types. Suppression of RPIA inhibits proliferation only in p16-low cells by inducing senescence both in vitro and in vivo. These data reveal the molecular basis whereby p16 loss modulates pro-tumorigenic metabolism through mTORC1-mediated upregulation of nucleotide synthesis and reveals a metabolic vulnerability of p16-null cancer cells.
Sigma-Aldrich
Uridine 5′-diphosphate disodium salt hydrate, ≥96.0% (HPLC)
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, 99%
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Charcoal Stripped, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial