Merck
Cbl interacts with multiple E2s in vitro and in cells.
PloS one (2019-05-24)
Mariya S Liyasova, Ke Ma, Donna Voeller, Philip E Ryan, Jinqiu Chen, Rachel E Klevit, Stanley Lipkowitz
ABSTRACT
Many receptor tyrosine kinases (RTKs, such as EGFR, MET) are negatively regulated by ubiquitination and degradation mediated by Cbl proteins, a family of RING finger (RF) ubiquitin ligases (E3s). Loss of Cbl protein function is associated with malignant transformation driven by increased RTK activity. RF E3s, such as the Cbl proteins, interact with a ubiquitin-conjugating enzyme (E2) to confer specificity to the ubiquitination process and direct the transfer of ubiquitin from the E2 to one or more lysines on the target proteins. Using in vitro E3 assays and yeast two-hybrid screens, we found that Ube2d, Ube2e families, Ube2n/2v1, and Ube2w catalyze autoubiquitination of the Cbl protein and Ube2d2, Ube2e1, and Ube 2n/2v1 catalyze Cbl-mediated substrate ubiquitination of the EGFR and SYK. Phosphorylation of the Cbl protein by by Src resulted in increased E3 activity compared to unphosphorylated cbl or Cbl containing a phosphomimetic Y371E mutation. Ubiquitin chain formation depended on the E2 tested with Cbl with Ube2d2 forming both K48 and K63 linked chains, Ube2n/2v1 forming only K63 linked chains, and Ube2w inducing monoubiquitination. In cells, the Ube2d family, Ube2e family, and Ube2n/2v1 contributed to EGFR ubiquitination. Our data suggest that multiple E2s can interact with Cbl and modulate its E3 activity in vitro and in cells.
Sigma-Aldrich
Anti-Ubiquitin Antibody, Lys63-Specific, clone Apu3, rabbit monoclonal, clone Apu3, from rabbit
Sigma-Aldrich
Anti-Src Antibody, clone N6L, rabbit monoclonal, culture supernatant, clone N6L, from rabbit
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid