Merck
  • Pex3 and Atg37 compete to regulate the interaction between the pexophagy receptor, Atg30, and the Hrr25 kinase.
Pex3 and Atg37 compete to regulate the interaction between the pexophagy receptor, Atg30, and the Hrr25 kinase.
Autophagy (2017-12-21)
Katarzyna Zientara-Rytter, Katharine Ozeki, Taras Y Nazarko, Suresh Subramani
ABSTRACT
Macroautophagy/autophagy is a highly conserved process in which subcellular components destined for degradation are sequestered within autophagosomes. The selectivity of autophagy is determined by autophagy receptors, such as Pichia pastoris Atg30 (autophagy-related 30), which controls the selective degradation of peroxisomes (pexophagy) through the assembly of a receptor-protein complex (RPC). Previously, we proved that the peroxisomal acyl-CoA-binding protein, Atg37, and the highly conserved peroxin, Pex3, are required for RPC formation and efficient pexophagy. Here, we describe how Atg37 and Pex3 regulate the assembly and activation of the pexophagic RPC. We demonstrate that Atg30 requires both Atg37 and Pex3 to recruit Atg8 and Atg11 to the pexophagic RPC, because Atg37 depends on Pex3 for its localization at the peroxisomal membrane. We establish that due to close proximity of Atg37- and Pex3-binding sites in the middle domain of Atg30, the binding of these proteins to Atg30 is mutually exclusive within this region. We also show that direct binding of Pex3 and Atg37 to Atg30 regulates its phosphorylation by the Hrr25 kinase, negatively and positively, respectively. Based on these results we present a model that clarifies the assembly and activation of the pexophagic RPC through the phosphoregulation of Atg30.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Calmodulin Antibody, Upstate®, from mouse