Merck
  • N⁶-Methyladenosine Landscape of Glioma Stem-Like Cells: METTL3 Is Essential for the Expression of Actively Transcribed Genes and Sustenance of the Oncogenic Signaling.
N⁶-Methyladenosine Landscape of Glioma Stem-Like Cells: METTL3 Is Essential for the Expression of Actively Transcribed Genes and Sustenance of the Oncogenic Signaling.
Genes (2019-02-20)
Abhirami Visvanathan, Vikas Patil, Shibla Abdulla, Jörg D Hoheisel, Kumaravel Somasundaram
ABSTRACT
Despite recent advances in N⁶-methyladenosine (m⁶A) biology, the regulation of crucial RNA processing steps by the RNA methyltransferase-like 3 (METTL3) in glioma stem-like cells (GSCs) remains obscure. An integrated analysis of m⁶A-RIP (RNA immunoprecipitation) and total RNA-Seq of METTL3-silenced GSCs identified that m⁶A modification in GSCs is principally carried out by METTL3. The m⁶A-modified transcripts showed higher abundance compared to non-modified transcripts. Further, we showed that the METTL3 is essential for the expression of GSC-specific actively transcribed genes. Silencing METTL3 resulted in the elevation of several aberrant alternative splicing events. We also found that putative m⁶A reader proteins play a key role in the RNA stabilization function of METTL3. METTL3 altered A-to-I and C-to-U RNA editing events by differentially regulating RNA editing enzymes ADAR and APOBEC3A. Similar to protein-coding genes, lincRNAs (long intergenic non-coding RNAs) with m⁶A marks showed METTL3-dependent high expression. m⁶A modification of 3'UTRs appeared to result in a conformation-dependent hindrance to miRNA binding to their targets. The integrated analysis of the m⁶A regulome in METTL3-silenced GSCs showed global disruption in tumorigenic pathways that are indispensable for GSC maintenance and glioma progression. We conclude that METTL3 plays a vital role in many steps of RNA processing and orchestrates successful execution of oncogenic pathways in GSCs.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%