Merck
  • Lysine trimethylation regulates 78-kDa glucose-regulated protein proteostasis during endoplasmic reticulum stress.
Lysine trimethylation regulates 78-kDa glucose-regulated protein proteostasis during endoplasmic reticulum stress.
The Journal of biological chemistry (2017-09-16)
Jonas Sieber, Nicolas Wieder, Mauricio Ostrosky-Frid, Moran Dvela-Levitt, Ozan Aygün, Namrata D Udeshi, Steven A Carr, Anna Greka
ABSTRACT
The up-regulation of chaperones such as the 78-kDa glucose-regulated protein (GRP78, also referred to as BiP or HSPA5) is part of the adaptive cellular response to endoplasmic reticulum (ER) stress. GRP78 is widely used as a marker of the unfolded protein response, associated with sustained ER stress. Here we report the discovery of a proteostatic mechanism involving GRP78 trimethylation in the context of ER stress. Using mass spectrometry-based proteomics, we identified two GRP78 fractions, one homeostatic and one induced by ER stress. ER stress leads to de novo biosynthesis of non-trimethylated GRP78, whereas homeostatic, METTL21A-dependent lysine 585-trimethylated GRP78 is reduced. This proteostatic mechanism, dependent on the posttranslational modification of GRP78, allows cells to differentially regulate specific protein abundance during cellular stress.
Sigma-Aldrich
Anti-HSPA5 Antibody, clone 4E3, ascites fluid, clone 4E3, from mouse