Merck
  • Developmental expression of the dynamin family of mechanoenzymes in the mouse epididymis.
Developmental expression of the dynamin family of mechanoenzymes in the mouse epididymis.
Biology of reproduction (2017-04-11)
Wei Zhou, Geoffry N De Iuliis, Adrian P Turner, Andrew T Reid, Amanda L Anderson, Adam McCluskey, Eileen A McLaughlin, Brett Nixon
ABSTRACT
The mammalian epididymis is an exceptionally long ductal system tasked with the provision of one of the most complex intraluminal fluids found in any exocrine gland. This specialized milieu is continuously modified by the combined secretory and absorptive of the surrounding epithelium and thus finely tuned for its essential roles in promoting sperm maturation and storage. While considerable effort has been focused on defining the composition of the epididymal fluid, relatively less is known about the intracellular trafficking machinery that regulates this luminal environment. Here, we characterize the ontogeny of expression of a master regulator of this machinery, the dynamin family of mechanoenzymes. Our data show that canonical dynamin isoforms were abundantly expressed in the juvenile mouse epididymis. However, in peripubertal and adult animals dynamin takes on a heterogeneous pattern of expression such that the different isoforms displayed both cell- and segment-specific localization. Thus, dynamin 1 and 3 were predominately localized in the distal epididymal segments (corpus and cauda), where they were found within clear and principal cells, respectively. In contrast, dynamin 2 was expressed throughout the epididymis, but localized to the Golgi apparatus of the principal cells in the proximal (caput) segment and the luminal border of these cells in more distal segments. These dynamin isoforms are therefore ideally positioned to play complementary, nonredundant roles in the regulation of the epididymal milieu. In support of this hypothesis, selective inhibition of dynamin altered the profile of proteins secreted from an immortalized caput epididymal cell line.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2