Merck
  • Biperiden and mepazine effectively inhibit MALT1 activity and tumor growth in pancreatic cancer.
Biperiden and mepazine effectively inhibit MALT1 activity and tumor growth in pancreatic cancer.
International journal of cancer (2019-07-11)
Leonie Konczalla, Daniel R Perez, Nadine Wenzel, Gerrit Wolters-Eisfeld, Clarissa Klemp, Johanna Lüddeke, Annika Wolski, Dirk Landschulze, Chris Meier, Anika Buchholz, Dichao Yao, Bianca T Hofmann, Julia K Graß, Sarah L Spriestersbach, Katharina Grupp, Udo Schumacher, Christian Betzel, Svetlana Kapis, Theresa Nuguid, Pablo Steinberg, Klaus Püschel, Guido Sauter, Maximillian Bockhorn, Faik G Uzunoglu, Jakob R Izbicki, Cenap Güngör, Alexander T El Gammal
ABSTRACT
MALT1 is a key mediator of NF-κB signaling and a main driver of B-cell lymphomas. Remarkably, MALT1 is expressed in the majority of pancreatic ductal adenocarcinomas (PDACs) as well, but absent from normal exocrine pancreatic tissue. Following, MALT1 shows off to be a specific target in cancer cells of PDAC without affecting regular pancreatic cells. Therefore, we studied the impact of pharmacological MALT1 inhibition in pancreatic cancer and showed promising effects on tumor progression. Mepazine (Mep), a phenothiazine derivative, is a known potent MALT1 inhibitor. Newly, we described that biperiden (Bip) is a potent MALT1 inhibitor with even less pharmacological side effects. Thus, Bip is a promising drug leading to reduced proliferation and increased apoptosis in PDAC cells in vitro and in vivo. By compromising MALT1 activity, nuclear translocation of c-Rel is prevented. c-Rel is critical for NF-κB-dependent inhibition of apoptosis. Hence, off-label use of Bip or Mep represents a promising new therapeutic approach to PDAC treatment. Regularly, the Anticholinergicum Bip is used to treat neurological side effects of Phenothiazines, like extrapyramidal symptoms.
Sigma-Aldrich
Fluoroshield with DAPI, liquid
Sigma-Aldrich
MALT1 Inhibitor II, Mepazine Hydrochloride
Sigma-Aldrich
MISSION® esiRNA, targeting human MALT1