Merck
  • DDX19 Inhibits Type I Interferon Production by Disrupting TBK1-IKKε-IRF3 Interactions and Promoting TBK1 and IKKε Degradation.
DDX19 Inhibits Type I Interferon Production by Disrupting TBK1-IKKε-IRF3 Interactions and Promoting TBK1 and IKKε Degradation.
Cell reports (2019-01-31)
Kunli Zhang, Yuanfeng Zhang, Juan Xue, Qingwen Meng, Hongyang Liu, Caihong Bi, Changyao Li, Liang Hu, Huibin Yu, Tao Xiong, Yuying Yang, Shangjin Cui, Zhigao Bu, Xijun He, Jiangnan Li, Li Huang, Changjiang Weng
ABSTRACT
DExD/H-box helicase members are key receptors for recognizing viral nucleic acids, and they regulate retinoic acid-inducible gene I (RIG-I)-like receptor (RLR)-mediated type I interferon (IFN) production. Here, we report that the DExD/H-box helicase family member DExD/H-box RNA helicase 19 (DDX19) is a negative regulator of type I IFN production. Ectopic expression of DDX19 suppressed poly(I:C) (polyinosinic-polycytidylic acid)- and Sendai-virus-induced type I IFN production, whereas knockdown of DDX19 expression enhanced type I IFN production. Mechanistically, DDX19 inhibited TANK-binds kinase 1 (TBK1)- and inhibitor-κb kinase ε (IKKε)-mediated phosphorylation of interferon regulatory factor 3 (IRF3) by disrupting the interaction between TBK1 or IKKε and IRF3. Additionally, DDX19 recruited Lamtor2 and then formed the TBK1-IKKε-Lamtor2-DDX19-IRF3 complex to suppress IFN production by promoting TBK1 and IKKε degradation. We generated Ddx19 knockout mice using transcription activator-like effector nucleases (TALENs) and found that Ddx19 deficiency in vivo augmented type I IFN production, resulting in suppression of encephalomyocarditis virus replication. These data show that DDX19 is an important negative regulator of RLR-mediated type I IFN production.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody