Merck
  • UPARANT is an effective antiangiogenic agent in a mouse model of rubeosis iridis.
UPARANT is an effective antiangiogenic agent in a mouse model of rubeosis iridis.
Journal of molecular medicine (Berlin, Germany) (2019-06-28)
Filippo Locri, Massimo Dal Monte, Monica Aronsson, Maurizio Cammalleri, Mario De Rosa, Vincenzo Pavone, Anders Kvanta, Paola Bagnoli, Helder André
ABSTRACT
Puncture-induced iris neovascularization (rubeosis iridis; RI) in mice is associated with upregulation of extracellular matrix (ECM) degradation and inflammatory factors. The anti-angiogenic and anti-inflammatory efficacy of UPARANT in reducing RI was determined by noninvasive, in vivo iris vascular densitometry, and confirmed in vitro by quantitative vascular-specific immunostaining. Intravitreal administration of UPARANT successfully and rapidly reduced RI to non-induced control levels. Molecular analysis revealed that UPARANT inhibits formyl peptide receptors through a predominantly anti-inflammatory response, accompanied with a significant reduction in ECM degradation and inflammation markers. Similar results were observed with UPARANT administered systemically by subcutaneous injection. These data suggest that the tetrapeptide UPARANT is an effective anti-angiogenic agent for the treatment of RI, both by local and systemic administrations. The effectiveness of UPARANT in reducing RI in a model independent of the canonical vascular endothelial growth factor (VEGF) proposes an alternative for patients that do not respond to anti-VEGF treatments, which could improve treatment in proliferative ocular diseases. KEY MESSAGES: UPARANT is effective in the treatment of rubeosis iridis, both by local and systemic administrations. UPARANT can reduce VEGF-independent neovascularization.
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution