Merck
  • Minicircle HBV cccDNA with a Gaussia luciferase reporter for investigating HBV cccDNA biology and developing cccDNA-targeting drugs.
Minicircle HBV cccDNA with a Gaussia luciferase reporter for investigating HBV cccDNA biology and developing cccDNA-targeting drugs.
Scientific reports (2016-11-08)
Feng Li, Liang Cheng, Christopher M Murphy, Natalia J Reszka-Blanco, Yaxu Wu, Liqun Chi, Jianming Hu, Lishan Su
ABSTRACT
Chronic Hepatitis B Virus (HBV) infection is generally not curable with current anti-viral drugs. Virus rebounds after stopping treatment from the stable HBV covalently-closed-circular DNA (cccDNA). The development of drugs that directly target cccDNA is hampered by the lack of robust HBV cccDNA models. We report here a novel HBV cccDNA technology that will meet the need. We engineered a minicircle HBV cccDNA with a Gaussia Luciferase reporter (mcHBV-GLuc cccDNA), which serves as a surrogate to measure cccDNA activity. The mcHBV-GLuc cccDNA was easily produced in bacteria, and it formed minichromosomes as HBV cccDNA episome DNA does when it was transfected into human hepatocytes. Compared to non-HBV minicircle plasmids, mcHBV-GLuc cccDNA showed persistent HBV-GLuc activity and HBx-dependent gene expression. Importantly, the mcHBV-GLuc cccDNA showed resistance to interferons (IFN) treatment, indicating its unique similarity to HBV cccDNA that is usually resistant to long-term IFN treatment in chronic HBV patients. Most importantly, GLuc illuminates cccDNA as a surrogate of cccDNA activity, providing a very sensitive and quick method to detect trace amount of cccDNA. The mcHBV-GLuc cccDNA model is independent of HBV infection, and will be valuable for investigating HBV cccDNA biology and for developing cccDNA-targeting drugs.
Sigma-Aldrich
ChIPAb+ HDAC1 - ChIP Validated Antibody and Primer Set, culture supernatant, from mouse
Sigma-Aldrich
ChIPAb+ Histone H2B - ChIP Validated Antibody and Primer Set, from rabbit
Sigma-Aldrich
ChIPAb+ PCAF Antibody
Sigma-Aldrich
ChIPAb+ Histone H4 - ChIP Validated Antibody and Primer Set, rabbit monoclonal, from rabbit
Sigma-Aldrich
ChIPAb+ Histone H3 (CT) - ChIP Validated Antibody and Primer Set, rabbit monoclonal, culture supernatant, from rabbit