Merck
  • ROS-Mediated 15-Hydroxyprostaglandin Dehydrogenase Degradation via Cysteine Oxidation Promotes NAD+-Mediated Epithelial-Mesenchymal Transition.
ROS-Mediated 15-Hydroxyprostaglandin Dehydrogenase Degradation via Cysteine Oxidation Promotes NAD+-Mediated Epithelial-Mesenchymal Transition.
Cell chemical biology (2018-01-09)
Weixuan Wang, Yadong Hu, Xiaofei Wang, Qingtao Wang, Haiteng Deng
ABSTRACT
Nicotinamide adenine dinucleotide (NAD) levels decrease with aging as a result of aging-associated CD38 upregulation. Here, we established a cell model with decreased cellular NAD levels by overexpressing CD38 or treating cells with FK866, an inhibitor of nicotinamide phosphoribosyltransferase. We revealed that decreased NAD triggered reactive oxygen species (ROS)-mediated degradation of 15-hydroxyprostaglandin dehydrogenase (15-PGDH), which drove cells to undergo epithelial-mesenchymal transition (EMT). Moreover, we showed that oxidation of the Cys44 residue to sulfonic acid in 15-PGDH led to its degradation via non-canonical ubiquitination-proteasome and autophagy pathways. Mutation of Cys44 to alanine abolished ROS-induced 15-PGDH degradation. We demonstrated that 15-PGDH silencing promoted EMT, whereas supplementation with NAD precursors increased NAD and 15-PGDH stability, and reversed the EMT process. Taken together, these results suggest that declining NAD levels contribute to age-dependent increases in cancer incidence, and repletion of NAD precursors is beneficial for increasing 15-PGDH expression.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-Cytokeratin 18 Antibody, clone RCK106, clone RCK106, Chemicon®, from mouse