Merck
  • Amyloid β-dependent mitochondrial toxicity in mouse microglia requires P2X7 receptor expression and is prevented by nimodipine.
Amyloid β-dependent mitochondrial toxicity in mouse microglia requires P2X7 receptor expression and is prevented by nimodipine.
Scientific reports (2019-04-26)
Paola Chiozzi, Alba Clara Sarti, Juana M Sanz, Anna Lisa Giuliani, Elena Adinolfi, Valentina Vultaggio-Poma, Simonetta Falzoni, Francesco Di Virgilio
ABSTRACT
Previous data from our laboratory show that expression of the P2X7 receptor (P2X7R) is needed for amyloid β (Aβ)-stimulated microglia activation and IL-1β release in vitro and in vivo. We also showed that Aβ-dependent stimulation is inhibited by the dihydropyridine nimodipine at an intracellular site distal to the P2X7R. In the present study, we used the N13 microglia cell line and mouse primary microglia from wt and P2rx7-deleted mice to test the effect of nimodipine on amyloid β (Aβ)-dependent NLRP3 inflammasome expression and function, and on mitochondrial energy metabolism. Our data show that in microglia Aβ causes P2X7R-dependent a) NFκB activation; b) NLRP3 inflammasome expression and function; c) mitochondria toxicity; and these changes are fully inhibited by nimodipine. Our study shows that nimodipine is a powerful blocker of cell damage caused by monomeric and oligomeric Aβ, points to the mitochondria as a crucial target, and underlines the permissive role of the P2X7R.