Merck
  • OTU deubiquitinase 4 is silenced and radiosensitizes non-small cell lung cancer cells via inhibiting DNA repair.
OTU deubiquitinase 4 is silenced and radiosensitizes non-small cell lung cancer cells via inhibiting DNA repair.
Cancer cell international (2019-04-24)
Zhiqiang Wu, Minghan Qiu, Yu Guo, Jinlin Zhao, Zhuang Liu, Hui Wang, Maobin Meng, Zhiyong Yuan, Zeyun Mi
ABSTRACT
Radiotherapy is becoming one major therapeutics for non-small cell lung cancer (NSCLC). Identifying novel radiosensitizers will greatly increase the efficacy of radiotherapy and benefit more patients. OTU deubiquitinase 4 (OTUD4) has been reported involved in DNA damage repair pathways and could be a potential target for chemotherapy therapy. This study aimed to investigate the roles of OTUD4 in regulation of radiosensitivity of NSCLC via modulating DNA repair. The expression of OTUD4, γ-H2Ax and ATM/CHK2/p53 pathway-related signaling molecules were detected by Western blotting and QRT-PCR. The methylation of OTUD4 promoter was investigated by 5-aza-deoxycytidine treatment, methylation-specific PCR and bisulfite genomic sequencing assays. Radiosensitivity was assessed by the clonogenic formation assay. Cell cycle, cell apoptosis were analyzed by flow cytometry. DNA damage and repair were determined by comet assay, γ-H2Ax foci staining and flow cytometry. OTUD4 is dramatically downregulated in NSCLC and its downregulation significantly correlates with poor prognosis of NSCLC patients. Promoter hypermethylation is responsible for the loss of OTUD4 expression in NSCLC cells. Overexpression of OTUD4 increases radiosensitivity of NSCLC cells exhibiting as impaired clonogenic formation ability, enhanced cell cycle arrest and increased cell apoptosis. Moreover, molecular mechanism study reveals that OTUD4 radiosensitizs NSCLC cells via ATM/CHK2/P53 signaling and inhibiting homology-directed repair of DNA double strand breaks induced by ionizing radiation. This study uncovers a tumor-suppressing role of OTUD4 and that OTUD4 is a potential radiosensitizer for NSCLC.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-OTUD4 Antibody, from rabbit, purified by affinity chromatography