Merck
E3 ubiquitin ligase RNF123 targets lamin B1 and lamin-binding proteins.
The FEBS journal (2018-04-21)
Richa Khanna, Vidhya Krishnamoorthy, Veena K Parnaik
ABSTRACT
Lamins are key nuclear proteins which are important for maintaining nuclear structure and function. Mutations in lamins cause a spectrum of genetic diseases termed as laminopathies. RING finger containing E3 ubiquitin ligase, RNF123, is transcriptionally upregulated in cells expressing rod domain lamin A mutations. However, the functional relevance of RNF123 in laminopathic cells is not clear. Using a mass spectrometry-based approach, we identified lamins and lamin-binding proteins retinoblastoma protein (pRb), lamina-associated polypeptide 2α (LAP2α), and emerin as RNF123-interacting proteins. We determined that RNF123 mediated the ubiquitination of these proteins and caused the proteasomal degradation of pRb, LAP2α, and lamin B1. Furthermore, these proteins were also targeted for proteasomal degradation in cells expressing lamin A rod domain mutants G232E, Q294P, and R386K. Overexpression of RNF123 resulted in delayed transit through the S-phase which was alleviated by coexpression of pRb or LAP2α. Our findings imply that RNF123-mediated ubiquitination of lamin-binding proteins may contribute to disease-causing mechanisms in laminopathies by depletion of key nuclear proteins and defects in cell cycle kinetics.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)