Merck
  • Elevated Peripheral Myelin Protein 22, Reduced Mitotic Potential, and Proteasome Impairment in Dermal Fibroblasts from Charcot-Marie-Tooth Disease Type 1A Patients.
Elevated Peripheral Myelin Protein 22, Reduced Mitotic Potential, and Proteasome Impairment in Dermal Fibroblasts from Charcot-Marie-Tooth Disease Type 1A Patients.
The American journal of pathology (2017-12-17)
Sooyeon Lee, Hannah Bazick, Vinita Chittoor-Vinod, Mohammed Omar Al Salihi, Guangbin Xia, Lucia Notterpek
ABSTRACT
A common form of hereditary autosomal dominant demyelinating neuropathy known as Charcot-Marie-Tooth disease type 1A (CMT1A) is linked with duplication of the peripheral myelin protein 22 (PMP22) gene. Although studies from animal models have led to better understanding of the pathobiology of these neuropathies, there continues to be a gap in the translation of findings from rodents to humans. Because PMP22 was originally identified in fibroblasts as growth arrest specific gene 3 (gas3) and is expressed broadly in the body, it was tested whether skin cells from neuropathic patients would display the cellular pathology observed in Schwann cells from rodent models. Dermal fibroblasts from two CMT1A pedigrees with confirmed PMP22 gene duplication were studied. Samples from age-matched non-neuropathic individuals were used as controls. CMT1A patient-derived cultures contain approximately 1.5-fold elevated levels of PMP22 mRNA, exhibit reduced mitotic potential, and display intracellular protein aggregates as compared to cells from unaffected individuals. The presence of cytosolic PMP22 coincides with a decrease in proteasome activity and an increase in autophagy-lysosomal proteins, including LC3-II and LAMP1. These results indicate that the abnormalities in the subcellular processing of excess PMP22 elicit a detectable response in human CMT1A fibroblasts, a phenotype that resembles Schwann cells from neuropathic mice. These findings support the use of human CMT1A fibroblasts as a platform for therapy testing.
Sigma-Aldrich
Anti-BrdU Antibody, clone 131-14871, clone 131-14871, Chemicon®, from mouse