Merck
  • Estradiol inhibits Th17 cell differentiation through inhibition of RORγT transcription by recruiting the ERα/REA complex to estrogen response elements of the RORγT promoter.
Estradiol inhibits Th17 cell differentiation through inhibition of RORγT transcription by recruiting the ERα/REA complex to estrogen response elements of the RORγT promoter.
Journal of immunology (Baltimore, Md. : 1950) (2015-03-15)
Rong-Yi Chen, Yi-Ming Fan, Qiuyang Zhang, Sen Liu, Qingli Li, Guo-Lin Ke, Chen Li, Zongbing You
ABSTRACT
The symptoms of vaginal candidiasis exacerbate in the second half of the menstrual cycle in premenopausal women when the serum estradiol level is elevated. Estradiol has been shown to inhibit Th17 differentiation and production of antifungal IL-17 cytokines. However, little is known about the mechanisms. In the present study, we used mouse splenocytes and found that estradiol inhibited Th17 differentiation through downregulation of Rorγt mRNA and protein expression. Estradiol activated estrogen receptor (ER)α to recruit repressor of estrogen receptor activity (REA) and form the ERα/REA complex. This complex bound to three estrogen response element (ERE) half-sites on the Rorγt promoter region to suppress Rorγt expression. Estradiol induced Rea mRNA and protein expression in mouse splenocytes. Using Rea small interfering RNA to knock down Rea expression enhanced Rorγt expression and Th17 differentiation. Alternatively, histone deacetylase 1 and 2 bound to the three ERE half-sites, independent of estradiol. Histone deacetylase inhibitor MS-275 dose- and time-dependently increased Rorγt expression and subsequently enhanced Th17 differentiation. In 15 healthy premenopausal women, high serum estradiol levels are correlated with low RORγT mRNA levels and high REA mRNA levels in the vaginal lavage. These results demonstrate that estradiol upregulates REA expression and recruits REA via ERα to the EREs on the RORγT promoter region, thus inhibiting RORγT expression and Th17 differentiation. This study suggests that the estradiol/ERα/REA axis may be a feasible target in the management of recurrent vaginal candidiasis.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
EZ-Magna ChIP® G - Chromatin Immunoprecipitation Kit, Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic G beads. Control primers included.
Sigma-Aldrich
Anti-REA Antibody, Upstate®, from rabbit
Sigma-Aldrich
ChIPAb+ Estrogen Receptor α - ChIP Validated Antibody and Primer Set, ascites fluid, from mouse