Merck
  • Fas activation induces renal tubular epithelial cell beta 8 integrin expression and function in the absence of apoptosis.
Fas activation induces renal tubular epithelial cell beta 8 integrin expression and function in the absence of apoptosis.
The Journal of biological chemistry (2002-09-27)
George Jarad, Bingcheng Wang, Shenaz Khan, Jay DeVore, Hui Miao, Karen Wu, Stephen L Nishimura, Barbara A Wible, Martha Konieczkowski, John R Sedor, Jeffrey R Schelling
ABSTRACT
Cell fate following Fas (CD95) ligand or agonistic anti-Fas antibody stimulation is determined by multiple factors, including Fas expression level, microdomain localization, and modulating cytokines. Highly expressed Fas clusters and activates a canonical apoptosis signaling pathway. In less susceptible cells, Fas transduces apoptosis-independent signals, which are not well defined, but have been linked to inflammation, angiogenesis, and fibrosis. To identify apoptosis-independent Fas pathways, cultured renal tubular epithelial cells were stimulated with agonistic anti-Fas antibodies under conditions that did not cause cell death. Analysis of filter cDNA microarrays revealed beta(8) integrin subunit mRNA induction in Fas-stimulated cells. beta(8) integrin mRNA expression increased within 3-6 h of Fas ligation due to enhanced mRNA stabilization, and mRNA increases were sustained for 48-72 h. Expression of plasma membrane beta(8) integrin, as well as its heterodimer partner alpha(v), was increased by Fas activation with a similar kinetic pattern. Fas-induced alpha(v)beta(8) expression correlated with increased migration to vitronectin, the ligand for alpha(v)beta(8). Results from studies with function-blocking antibodies against other alpha(v)beta integrins or suppression of beta(8) integrin expression by RNA interference demonstrated that induced beta(8) integrin expression mediated Fas-stimulated migration. We conclude that alpha(v)beta(8) integrin induction defines an unexpected role for Fas in cell migration, rather than as a cell death receptor.
Sigma-Aldrich
Anti-Integrin αVβ5 Antibody, clone P1F6, azide free, clone P1F6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, Azide Free, clone LM609, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin αVβ6 Antibody, clone 10D5, azide free, clone 10D5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin αV Antibody, extracellular, clone AV1, azide free, culture supernatant, clone AV1, Chemicon®