Merck
  • Transcriptomic Analysis of Single Isolated Myofibers Identifies miR-27a-3p and miR-142-3p as Regulators of Metabolism in Skeletal Muscle.
Transcriptomic Analysis of Single Isolated Myofibers Identifies miR-27a-3p and miR-142-3p as Regulators of Metabolism in Skeletal Muscle.
Cell reports (2019-03-28)
Francesco Chemello, Francesca Grespi, Alessandra Zulian, Pasqua Cancellara, Etienne Hebert-Chatelain, Paolo Martini, Camilla Bean, Enrico Alessio, Lisa Buson, Martina Bazzega, Andrea Armani, Marco Sandri, Ruggero Ferrazza, Paolo Laveder, Graziano Guella, Carlo Reggiani, Chiara Romualdi, Paolo Bernardi, Luca Scorrano, Stefano Cagnin, Gerolamo Lanfranchi
ABSTRACT
Skeletal muscle is composed of different myofiber types that preferentially use glucose or lipids for ATP production. How fuel preference is regulated in these post-mitotic cells is largely unknown, making this issue a key question in the fields of muscle and whole-body metabolism. Here, we show that microRNAs (miRNAs) play a role in defining myofiber metabolic profiles. mRNA and miRNA signatures of all myofiber types obtained at the single-cell level unveiled fiber-specific regulatory networks and identified two master miRNAs that coordinately control myofiber fuel preference and mitochondrial morphology. Our work provides a complete and integrated mouse myofiber type-specific catalog of gene and miRNA expression and establishes miR-27a-3p and miR-142-3p as regulators of lipid use in skeletal muscle.
Sigma-Aldrich
Complete Whole Transcriptome Amplification Kit, DNA polymerase included, Complete Kit with optimized enzyme to amplify total RNA in <4 hours, no 3′ bias