Merck
  • High expression of Sonic hedgehog in allergic airway epithelia contributes to goblet cell metaplasia.
High expression of Sonic hedgehog in allergic airway epithelia contributes to goblet cell metaplasia.
Mucosal immunology (2018-06-06)
Chengyun Xu, Chaochun Zou, Musaddique Hussain, Wei Shi, Yanan Shao, Ziyan Jiang, Xiling Wu, Meiping Lu, Junsong Wu, Qiangmin Xie, Yuehai Ke, Fanxin Long, Lanfang Tang, Ximei Wu
ABSTRACT
Sonic hedgehog (SHH) is abundantly expressed and critical for morphogenesis in embryonic lungs; however, SHH expression drops to a much lower level in mice from E17.5 and in humans from the 21st gestational week. We find that SHH expression is robustly upregulated in the airway epithelia of children with asthma or mouse models with allergic airway disease. Specifically, airway-specific SMO loss of function significantly suppresses allergen-induced goblet cell phenotypes, whereas an airway-specific SMO gain of function markedly enhances the goblet cell phenotypes in mouse models with allergic airway disease. Notably, intratracheal administration with SHH-neutralizing antibody or cyclopamine robustly attenuates goblet cell phenotypes in mouse models with allergic airway disease. Finally, we identify that Muc5AC gene encoding MUC5AC mucin serves as a direct target of GLI transcriptional factors in response to SHH, whereas the SAM-pointed domain-containing ETS transcription factor and Forkhead box A2, critical transcriptional factors for goblet cell phenotypes, both function as the effectors of GLIs in response to SHH stimulation. Together, the upregulation of SHH expression in allergic bronchial epithelia contributes to goblet cell metaplasia; thus, blockage of SHH signaling is a rational approach in a therapeutic intervention of epithelial remodeling in chronic airway diseases.
Sigma-Aldrich
Anti-Protein patched homolog 1 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Sonic Hedgehog Antibody, from rabbit, purified by affinity chromatography